Double Sided Mirrors
Supplier: United Scientific Supplies
Jump to:
À propos de cet article
Flexible mirrors for optical experiments.
- Acts as a concave or convex mirror
- Can be cut with scissors
You'll get lots of use out of these double-sided, flexible plastic mirrors that can be used for investigations, observations and creative activities involving reflection, symmetry and more. The mirrors can be cut with scissors.
3 Options Available Below
Product Details & Documents
Flexible mirrors for optical experiments.
- Acts as a concave or convex mirror
- Can be cut with scissors
You'll get lots of use out of these double-sided, flexible plastic mirrors that can be used for investigations, observations and creative activities involving reflection, symmetry and more. The mirrors can be cut with scissors.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: MSA2X3-F
- Supplier Number: MSA4X6-F
- Supplier Number: MSA8X11-F