Dot Blot Analysis Activity Kit
Catalogue # 470149-882
Fournisseur: G-Biosciences
Spécifications
- Description:Dot Blot Analysis Activity Kit
- Cat. no.:470149-882
- Supplier No.:BE-502
Spécifications
À propos de cet article
Study Immunodetection Techniques
The Dot Blot technique is used in clinical testing, forensic, and research laboratories to detect disease. In this hands-on lab activity, students will use Dot Blot procedure to screen test samples from a population and identify those who are carriers of a disease. Materials supplied for six student groups for up to five students each.