Document Analysis Lab Activity
Document Analysis Lab Activity
Supplier:   Kemtec
Document Analysis Lab Activity
Supplier:  Kemtec
Certificates
Document Analysis Lab Activity
Document Analysis Lab Activity
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Determine a document’s authenticity.
  • Grades 6-12
  • Analysis of secret messages using chemistry
Your students will be fascinated as they perform both physical and chemical analysis of handwriting materials found at a crime scene. The procedures they will follow cover checking for erasures, abnormal positioning of a signature, use of different inks, and differences in handwriting or typing. It comes with enough materials for six setups and an instruction manual. A UV lamp, required, is available separately.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • Document Analysis Kit
    6
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $528.60
    $528.60
    Kit of 1
       $528.60
    Supplier Number: 10-506
  • Analysis of Documents Kit Refill
    6
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $339.85
    $339.85
    Each
       $339.85
    Supplier Number: 10-506R
  • Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -