DNA Fingerprinting: A Simple Simulation
DNA Finger Printing: A Simple Simulation
Catalog # 470004-150
Supplier: Avantor
Jump to:
Product Details & Documents
A clear and easy way to show students the steps of DNA fingerprinting by simulation, this kit with accompanying DVD guides students through the step by step process: from gel to Southern blot to radioactive probe to fingerprint using everyday UPC codes.
- Grades 10 to 12
- Developed by Julie Bohrer
Students can then compare suspects with crime scene DNA.
Delivery information: Includes materials for 15 student groups with DVD and instructions.
Spécifications
- Désignation:DNA Finger Printing: A Simple Simulation