DNA DuraGel™ | Avantor
DNA DuraGel™
Supplier:   Edvotek
DNA DuraGel™
Supplier:  Edvotek
Certificates
DNA DuraGel™
2 Options Available Below
Product Details & Documents
Ideal for student skill practice.
  • Durable, reusable practice gels
  • Accurately replicate actual agarose gel
  • Detailed instructions included
DNA DuraGel™ gels are permanent polymer gels that allow students to practice the critically important skill of pipetting/gel loading. The clear, reusable gels are designed for the practice of loading 5 – 35 µL of samples. The gel grids are imprinted with a ruler for sizing DNA fragments. Also included are simulated FlashBlue™ and Ethidium Bromide gel images, ideal for representing how actual gels are stained with Methylene Blue and Ethidium Bromide. Pipetting DNA and biological reagents is a critical skill required for biotechnology and life science experiments. This DNA DuraGel™ is designed for proper student training, to teach techniques for pipetting samples for agarose gel electrophoresis.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • DNA DuraGel™ kit
    6
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $124.80
    $124.80
    Pack of 6
       $124.80
    Supplier Number: S-43
  • DNA DuraGel™ set of 2 gels
    2
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $77.40
    $77.40
    Pack of 2
       $77.40
    Supplier Number: S-43-20