Dissolved Oxygen
À propos de cet article
Enough materials for five groups working simultaneously.
- Contains enough materials for a total of 25 tests
- The kit will measure 0, 4, and 8 ppm
Provide your students with hands-on reinforcement of key concepts in water quality and water treatment with this test kit. Includes a teacher’s manual with lesson plans, background information, and reproducible student procedure and data sheets.
1 Options Available Below
Product Details & Documents
Enough materials for five groups working simultaneously.
- Contains enough materials for a total of 25 tests
- The kit will measure 0, 4, and 8 ppm
Provide your students with hands-on reinforcement of key concepts in water quality and water treatment with this test kit. Includes a teacher’s manual with lesson plans, background information, and reproducible student procedure and data sheets.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 3976A-H
- Discontinued-Supplier Number: -