Dissection Aprons | Avantor
Dissection Aprons
Supplier:   Ward's Science
Dissection Aprons
Supplier:  Ward's Science
Certificates
Dissection Aprons
Dissection Aprons
2 Options Available Below
Product Details & Documents
Protects and educates.
  • Protective and disposable
  • Diagrams on aprons help students dissect
Help your students protect their clothing from chemicals and stains and provide them with a dissection guide at the same time. Each 3 mil, polyethylene apron features a black and white illustration of a dissected and labeled fetal pig or frog printed on it. Each pattern comes in a perforated roll of 100 aprons.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Frog Anatomy Aprons
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $184.05
    $184.05
    Pack of 100
       $184.05
    Supplier Number: 420.6100.1
  • Fetal Pig Anatomy Aprons
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $184.05
    $184.05
    Pack of 100
       $184.05
    Supplier Number: 420.6110.1