Disposable Specimen Containers | Avantor
Disposable Specimen Containers
Supplier:   Dynalon
Disposable Specimen Containers
Supplier:  Dynalon
Certificates
Disposable Specimen Containers
1 Options Available Below
Product Details & Documents
Keep specimens organized and out of your way with these handy storage units.
Featuring high-density polyethylene construction and tight-fitting lids, these translucent containers are resistant to freezing and boiling, and are sturdy enough to be used multiple times.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Specimen Container, 0.91 L, Disposable
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1.95
    $1.95
    Liquidation
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $192.50
    $192.50
    Liquidation
    Each
       $1.95
    Case of 100
       $192.50
    Supplier Number: 453615
  • Specimen Container, 1.81 L, Disposable
    Discontinued
    Supplier Number: 453625