ContactUsLinkComponent
Overflow Can with Pour Spout
Catalog # 470133-930
Supplier:  GSC International
Overflow Can with Pour Spout
Catalog # 470133-930
Supplier:  GSC International
Supplier Number:  4-15002
Specifications
  • Description:
    Overflow Can with Pour Spout
  • Cat. no.:
    470133-930
  • Supplier No.:
    7-1502
Specifications
About this item
Great for displacement experiments and density measurements.
  • Made from seamless aluminum
  • Pour spout arm
  • 80 mm (3.14") in diameter