Disk Dispenser | Avantor
Disk Dispenser
Disk Dispenser
Catalog # 470111-914
Supplier:   BBL
Fournisseur #:  260457
Disk Dispenser
Catalog # 470111-914
Supplier:  BBL
Disk Dispenser
Disk Dispenser
Disk Dispenser
Price
$184.60
Log in pour la disponibilité
Each
Product Details & Documents
Dispenser holds a single cartridge of antibiotic disks and eliminates the need for the use of forceps.
Spécifications
  • Désignation:
    Disk Dispenser