Disinfectant Solution | Avantor
Disinfectant Solution
Disinfectant Solution, Laboratory, 100 mL Bottle, Laboratory
Catalog # 470024-690
Supplier:   Ward's Science
Fournisseur #:  BELOW
Disinfectant Solution
Catalog # 470024-690
Supplier:  Ward's Science
CAS Number:  
Disinfectant Solution
Disinfectant Solution
Disinfectant Solution, Laboratory, 100 mL Bottle, Laboratory
CAS Number:  
Price
$21.40
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For sterilizing plant materials prior to tissue culture.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Spécifications
Spécifications
  • Désignation:
    Disinfectant Solution, Laboratory, 100 mL Bottle
  • Grade:
    Laboratory