Didinium
Supplier: Ward's Science
Jump to:
À propos de cet article
Predatory. Feeds on Paramecium caudatum. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.
2 Options Available Below
Product Details & Documents
Predatory. Feeds on Paramecium caudatum. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.