Eisco® Stem Structure Models | Avantor
Eisco® Stem Structure Models
Supplier:   Wards
Eisco® Stem Structure Models
Supplier:  Wards
Certificates
Eisco® Stem Structure Models
Eisco® Stem Structure Models
2 Options Available Below
Product Details & Documents
These stem models are used to show common structures, tissues, and vascular bundles of monocot or dicot stems.
Each model is cut in transverse section to view and Mounted on a plastic base.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Parts
    Width
    Longueur
    Hauteur
    Availability
    Price
  • Dicot Stem
    1
    25 cm
    33 cm
    5 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $171.55
    $171.55
    Each
       $171.55
    Supplier Number: BM0012
  • Monocot Stem
    1
    25 cm
    33 cm
    5 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $171.55
    $171.55
    Each
       $171.55
    Supplier Number: BM0016