Eisco® Root Structure Models | Avantor
Eisco® Root Structure Models
Supplier:   Wards
Eisco® Root Structure Models
Supplier:  Wards
Certificates
Eisco® Root Structure Models
Eisco® Root Structure Models
2 Options Available Below
Product Details & Documents
A traverse section of a monocot or a dicot root illustrates the various tissues in vascular bundles.
Mounted on a plastic base, the model includes a key card for all numbered structures.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Parts
    Width
    Longueur
    Hauteur
    Availability
    Price
  • Dicot Root
    1
    25 cm
    33 cm
    5 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $167.10
    $167.10
    Each
       $167.10
    Supplier Number: BM0024
  • Monocot Root
    1
    25 cm
    33 cm
    5 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $166.80
    $166.80
    Each
       $166.80
    Supplier Number: BM0026