Eisco® Leaf Structure Models | Avantor
Eisco® Leaf Structure Models
Supplier:   Wards
Eisco® Leaf Structure Models
Supplier:  Wards
Certificates
Eisco® Leaf Structure Models
Eisco® Leaf Structure Models
2 Options Available Below
Product Details & Documents
Examine the various tissues in vascular bundles found in these vertical section of a dicot and Monocot leaves.
  • Models Mounted on Plastic Base
  • Key Card Included
  • Models Demonstrate Tissues in Vascular Bundles
The models are mounted on a plastic base.
Each model includes a key card for all numbered structures.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Parts
    Width
    Longueur
    Hauteur
    Availability
    Price
  • Dicot Leaf
    1
    25 cm
    33 cm
    7 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $167.10
    $167.10
    Each
       $167.10
    Supplier Number: BM0020
  • Monocot Leaf
    1
    25 cm
    33 cm
    5 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $166.80
    $166.80
    Each
       $166.80
    Supplier Number: BM0022