Dial Altimeter/Barometer
Dial Altimeter/Barometer
Catalog # 470018-694
Supplier: SUN COMPANY, INC.
Fournisseur #: 203-F
Jump to:
Product Details & Documents
Compact, 2-in-1 field tool and weather instrument.
- Aligned with GLOBE protocols
This great compass companion measures altitude from 0–15,000 feet and barometric pressure from 17–31" Hg. It is protected by a hardside case. Size: 2 3/4" x 3 1/2" x 1".
Spécifications
- Désignation:Dial Altimeter/Barometer