Determining Vitamin C Concentration Lab Activity | Avantor
Determining Vitamin C Concentration Lab Activity
WARD’S Determining Vitamin C Concentration Lab Activity
Catalog # 470004-922
Supplier:   Avantor
Determining Vitamin C Concentration Lab Activity
Catalog # 470004-922
Supplier:  Avantor
Determining Vitamin C Concentration Lab Activity
Determining Vitamin C Concentration Lab Activity
WARD’S Determining Vitamin C Concentration Lab Activity
Price
86,99 $
Log in pour la disponibilité
KIT(1items) Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal to determine the amount of Vitamin C in fruits and vegetables.
Scientists are discovering that vitamin C plays a major role in human physiology. It is a key component in the formation of collagen, bones, and teeth, and it aids in fighting infection. In this interesting activity, students first titrate a known, standard solution of vitamin C. They then prepare their own samples using common fruits and vegetables (fresh, canned, or frozen) that you supply. By comparing their results to the standard solution, students can quantify the amount of vitamin C present in their own samples. It includes enough materials for eight setups, a teacher’s guide, and student study guide copymaster.
Spécifications
  • Désignation:
    WARD’S Determining Vitamin C Concentration Lab Activity