Dessicator Plate
Supplier: SILICO & CHEMICO PORCELAIN SE
Jump to:
About this item
Dessicator Plate is reliable and cost effective.
- Sturdy and strong
- Glazed top surface
- Fragile
This durable porcelain plate is three feet to keep it steady.
1 Options Available Below
Product Details & Documents
Dessicator Plate is reliable and cost effective.
- Sturdy and strong
- Glazed top surface
- Fragile
This durable porcelain plate is three feet to keep it steady.
Product Family Options
Product Information
- DésignationHauteurØ trouDimensions de la plaqueØ ext.Nb de placesWeightAvailabilityPrice
- Scheibler-Form Desiccator Plates16 mm30 mm30 mm14 cm50.4500Discontinued-Supplier Number: J-274
- Dessicator Plate16 mm30 mm1–95 mm—33.2000Each$90.20Case of 8$651.85Supplier Number: J-272
- Scheibler-Form Desiccator Plates16 mm30 mm30 mm14 cm53.6500Discontinued-Supplier Number: J-274