Density Identification Set
Catalogue # 470149-492
Fournisseur: Shiv Dial Sud & Sons
Spécifications
- Description:Density Identification Set
- Cat. no.:470149-492
- Supplier No.:P10043A
Spécifications
À propos de cet article
The Density Identification Set is ideal for an investigation lab.
- Twelve Different Specimens to Test
- Aluminum Cylinders Included
- Specimens Delivered in Wood Storage Block
- Linseed Oil Coating Protects Block from Wet Specimens
- Ideal for Investigation Labs
This density-cylinder set consists of twelve half-inch dia. specimens of varying lengths and materials.
The specimens are supplied in an attractive wooden storage block. The block is protected from the wet specimens by a linseed oil coating.
Ordering information: The set includes cylinders of aluminum, brass, copper, Pyrex glass, hard rubber, Delrin, PVC, acrylic, Teflon, and Nylon plastics, and poplar and oak wood.
Packaging: One set includes 12 specimens.