Density Cylinder Set
Cylinder, Density Set, Set/4
Catalog # 470138-850
Supplier: Wards
Fournisseur #: PH0112F
Jump to:
Product Details & Documents
This Equal Mass Density Set is for specific gravity and density determinations.
- For Specific Gravity and Density Determinations
- Four Cylinders of Varying Volumes Included
- Width of All Cylinders: 16 mm dia.
- Consist of: Cu, Fe, Al, Zn
It's used to study the specific relationship between mass and density.
The set includes four cylinders with the same mass and diameter (16 mm) but varying volumes.
Spécifications
- Désignation:Cylinder, Density Set, Set/4