Demonstration Pulleys | Avantor
Demonstration Pulleys
Supplier:   GSC International
Demonstration Pulleys
Supplier:  GSC International
Certificates
Demonstration Pulleys
Demonstration Pulleys
2 Options Available Below
Product Details & Documents
Jumbo-Sized Pulleys are Designed for Classroom Demonstrations
These pulleys are open to make it easier for users to run pulley cords through them. The single, double, and triple pulleys are all 2 3/4" in diameter. Light-weight, low-friction, and deformation-resistant plastic ensures accurate results.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Demonstration Pulleys, Single
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9.50
    $9.50
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $86.90
    $86.90
    Each
       $9.50
    Pack of 10
       $86.90
    Supplier Number: 4217-00-10
  • Demonstration Pulleys, Double
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10.25
    $10.25
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $94.80
    $94.80
    Each
       $10.25
    Pack of 10
       $94.80
    Supplier Number: 4218-00-10
  • Demonstration Pulleys, Triple
    Discontinued
    Supplier Number: 4219-00-10