Deluxe Box Magnifier
470235-718
- Magnification:3×
- Description:Deluxe Box Magnifier
- Cat. no.:470235-718
Ideal for Observing Large Insects.
- Base size: 3 1/2" L x 3 1/2" W x 2 3/4" H
- Lens Ø: 2 1/4"
View specimens up close, then set them free. The base of the 3X magnifier covers live specimens for hands-free observation. Clear plastic.