Dehydrated Media
Dehydrated Media
Certificates
Dehydrated Media
8 Options Available Below
Product Details & Documents
Dehydrated Media is suitable for almost all educational applications.
  • For Use with Plates, Slants, and Broth Media
  • Instructions Included
  • Sealed Bottle
  • Expiration Date on Bottle
It arrives in a sealed bottle with an expiration date and complete instructions for preparation. Once items have shipped, they have three or more years before expiration.
Agar and components are used for plates or slants. Assuming a 20 mL fill for a 100 x 15 mm petri dish, each 114 g bottle will fill at least 100 dishes; each 454 g bottle will fill at least 400. Broth media are used for tube cultures. Assuming a 12 mL fill for a 20 x 75 mm tube, each 114 g bottle will fill at least 1000 tubes; each 454 g bottle will fill at least 2000.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Type de conditionnement
    Availability
    Price
  • Dehydrated Media Potato Dextrose Agar
    114 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $110.80
    $110.80
    Each
       $110.80
    Supplier Number: CD149A
  • Dehydrated Media Beef Extract (component)
    114 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $78.30
    $78.30
    Each
       $78.30
    Supplier Number: CD228A
  • Dehydrated Media Nutrient Agar
    454 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.80
    $218.80
    Each
       $218.80
    Supplier Number: LD145B
  • Dehydrated Media Nutrient Agar
    114 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $76.70
    $76.70
    Each
       $76.70
    Supplier Number: LD145A
  • Dehydrated Media Tryptone (component)
    114 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $62.50
    $62.50
    Each
       $62.50
    Supplier Number: CD351A
  • Dehydrated Media Yeast Extract (component)
    114 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.30
    $63.30
    Each
       $63.30
    Supplier Number: LD184A
  • Dehydrated Media Nutrient Broth
    114 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $77.00
    $77.00
    Each
       $77.00
    Supplier Number: CD146A
  • Dehydrated Media Tryptic Soy Agar
    114 g
    Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $48.60
    $48.60
    Each
       $48.60
    Supplier Number: LD100A