ContactUsLinkComponent
Damselfly, Adult
Catalogue # 470001-300
Fournisseur:  Avantor
undefined
Damselfly, Adult
Catalogue # 470001-300
Fournisseur:  Avantor
Spécifications
  • Description:
    Damselfly, Adult, Preserved, Jar of 10
  • Cat. no.:
    470001-300
  • Supplier No.:
    WARD470001-300
Spécifications
À propos de cet article
Ischnura sp. It has characteristic laterally projecting eyes and petiolate wings that fold above the body when at rest. Body size: 1 1/2".