Cylinder Guards
Cylinder Guards
Supplier:   Plasticoid
Cylinder Guards
Supplier:  Plasticoid
Certificates
Cylinder Guards
1 Options Available Below
Product Details & Documents
Black sponge rubber guards prevent breakage and rolling.
  • Hexagonal
  • 13 mm thick
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Cylinder Guards, For Cylinder Cap., 5 – 10 mL
    Discontinued
    Supplier Number: VCYCG1
  • Cylinder Guards, For Cylinder Cap., 25 – 50 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5.25
    $5.25
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $56.95
    $56.95
    Each
       $5.25
    Pack of 12
       $56.95
    Supplier Number: VCYCG2
  • Cylinder Guards, For Cylinder Cap., 100 mL
    Discontinued
    Supplier Number: VCYCG3