Cycloid, Scale Slide
Cycloid, Scale, Whole mount
Catalog # 470181-718
Supplier: Avantor
Jump to:
Product Details & Documents
Scale Type of Fish
- Identifying features clearly distinguishable
- Rigorous quality control standards
Stained. Isolated scale showing characteristic smooth curve of distal border.
Spécifications
- Désignation:Cycloid, Scale
- Orientation:Whole mount