Cuvette Rack
Cuvette Rack
Catalog # 470149-208
Supplier: Ward's Science
Supplier #: 260010
Jump to:
Product Details & Documents
Accommodate Most Square Cuvettes.
- Reduced risk of spilling
- Avoids risk of confusion with individually numbered spots
- Firmly holds up to 20 individual cuvettes
Specifications
- Désignation:Cuvette Rack