Ctenoid, Scale Slide
Ctenoid, Scale, Whole mount
Catalog # 470181-720
Supplier: Avantor
Jump to:
Product Details & Documents
Common Bony Fish Scale
- Identifying features clearly distinguishable
- Rigorous quality control standards
Stained, isolated scale showing characteristic spines of distal border.
Spécifications
- Désignation:Ctenoid, Scale
- Orientation:Whole mount