Crystal Models
Crystal Models
Crystal Models
Certificates
Crystal Models
3 Options Available Below
Product Details & Documents
Gorgeous Visual of Crystals
  • Introduce Crystal Lattices to your Students
  • Available in Three Types
These beautiful large crystal models enable you to introduce crystal lattices and unit cells to your classroom. Not only are they durable and provide for a great visual, they are also mounted permanently onto a clear acrylic base to minimize the possibility of lost parts.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Sodium Chloride
    Log in for availability
    Log in for availability
    Each
    Each - $61.45
    $61.45
    Clearance
    Each
       $61.45
    Supplier Number: CMSSCL
  • Graphite
    Log in for availability
    Log in for availability
    Each
    Each - $66.85
    $66.85
    Clearance
    Each
       $66.85
    Supplier Number: CMSGRP
  • Diamond
    Log in for availability
    Log in for availability
    Each
    Each - $67.40
    $67.40
    Clearance
    Each
       $67.40
    Supplier Number: CMSDMD