Crystal Form Topic Set | Avantor
Crystal Form Topic Set
Crystal form topic set
Catalog # 470025-344
Supplier:   Avantor
Crystal Form Topic Set
Catalog # 470025-344
Supplier:  Avantor
Crystal Form Topic Set
Crystal Form Topic Set
Crystal form topic set
Price
$118.99
Log in pour la disponibilité
Each
Product Details & Documents
Geometric Shapes and Structures of Crystalline Minerals.
Students can easily learn the primary crystal structures with the samples and activities contained in our complete GEO-logic study set. A teacher's guide and student copymasters are included for the following activities: crystal models, crystal form and habit—measuring crystal faces, and identifying crystal systems. The set is housed in a compartmented collection box.
Contents: 12 numbered samples, 1×1"; magnifier; protractor; ruler Isometric: grossular, pyrite (2) Hexagonal: apatite, corundum, dolomite Tetragonal: vesuvianite, zircon Orthorhombic: aragonite Monoclinic: orthoclase, staurolite Triclinic: microcline.
GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides. The GEO-logic system provides a modular approach to teaching and allows you to address broad curriculum needs for the study of minerals and rocks; natural resources and energy; and fossils and earth history.
The two types of GEO-logic teaching collections include:
• A Reference Set, which provides an overview of all topics and comes with a teacher's guide
• Activity Sets, which provide greater depth in an individual subject area and include both a teacher’s guide and student copymasters.
The GEO-logic system is:
• Developed jointly by experienced educators and WARD’S staff geologists
• Designed to save valuable planning and preparation time
• Created from quality teaching materials for hands-on learning
• Classroom tested for dependability.
Each GEO-logic collection includes:
• Individually numbered specimens chosen for their quality and character
• Activity guides with learning objectives and hands-on activities for your students to perform
• An attractive, durable compartmented box for easy use and storage.
Spécifications
  • Désignation:
    Crystal form topic set