Simulaids® CPR Torso
Lifelike anatomical landmarks such as a carotid pulse, sternum, ribcage, and sub sternal notch provide realistic CPR practice.
- Lightweight, easy-to-transport torso
- Realistic anatomical landmarks
- Easy to use
- Unit includes a carrying case
The torso features an airway ball and valve, which allows the lungs to inflate only if the head is extended and accurately positioned, so students learn and practice the proper procedure. The lightweight model is easy to transport and includes a soft carrying case with kneeling pads and simple-to-replace mouth/nose pieces.
The version with electronics has all the same features and also includes a light controller that confirms correct hand placement, ventilation, and compression.
Lifelike anatomical landmarks such as a carotid pulse, sternum, ribcage, and sub sternal notch provide realistic CPR practice.
- Lightweight, easy-to-transport torso
- Realistic anatomical landmarks
- Easy to use
- Unit includes a carrying case
The torso features an airway ball and valve, which allows the lungs to inflate only if the head is extended and accurately positioned, so students learn and practice the proper procedure. The lightweight model is easy to transport and includes a soft carrying case with kneeling pads and simple-to-replace mouth/nose pieces.
The version with electronics has all the same features and also includes a light controller that confirms correct hand placement, ventilation, and compression.
Product Family Options
Trainers
- Désignation
- : 100-2300
Replacement Materials
- Désignation
- : 100-2023
- Replacement Airways (Pk/10)-: -