Cork Slide
Cork, Section, Safranin O
Catalog # 470177-680
Supplier: Avantor
Jump to:
Product Details & Documents
Buoyant plant material
- Identifying features clearly distinguishable
- Rigorous quality control standards
General structure of the outer tissue of Spanish oak. The walls around air spaces reminded Robert Hooke of small rooms in a monastery called “cells”.
Spécifications
- Désignation:Cork
- Stain:Safranin O
- Orientation:Section