Ward's® Freshwater Copepods | Avantor
Ward's® Freshwater Copepods
Copepods, Freshwater, 35
Catalog # 470176-228
Supplier:   Ward's Science
Fournisseur #:  876020
Ward's® Freshwater Copepods
Catalog # 470176-228
Supplier:  Ward's Science
Ward's® Freshwater Copepods
Ward's® Freshwater Copepods
Copepods, Freshwater, 35
Price
$22.05
Log in pour la disponibilité
Each
Product Details & Documents
The culture supplied includes copepods in various stages of development.
Cyclops or similar genus. Copepods molt numerous times in the nauplius and copepodid stages before reaching adulthood.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Spécifications
  • Désignation:
    Copepods, Freshwater
  • No. of Students:
    35