Copepod Slide
Copepod, Whole mount, Schneider's and Fast Green
Catalog # 470181-592
Supplier: Avantor
Jump to:
Product Details & Documents
Major Component of Zooplankton
- Identifying features clearly distinguishable
- Rigorous quality control standards
Demonstrates the general body structure of a copepod.
Spécifications
- Désignation:Copepod
- Stain:Schneider's and Fast Green
- Orientation:Whole mount