Coolant Trap | Avantor
Coolant Trap
Coolant Trap
Catalog # 470022-252
Supplier:   GM DIECRON, INC.
Fournisseur #:  28-1921
Coolant Trap
Catalog # 470022-252
Supplier:  GM DIECRON, INC.
Coolant Trap
Coolant Trap
Price
$2,595.45
Log in pour la disponibilité
Each
Product Details & Documents
Extends pump life.
  • Keeps coolant and other debris from entering the vacuum pump
  • Suitable for all thin section saws/grinders sold by WARD’S
Spécifications
  • Désignation:
    Coolant Trap