Convex/Concave Mirrors
Convex/concave mirrors, glass, 200 mm, 75 mm
Catalog # 470006-844
Supplier: GSC International
Fournisseur #: 4-1301N
Jump to:
Product Details & Documents
Economical Convex/Concave Mirrors are dual purpose.
- Dual-Purpose
- Smooth Edges
- Circular Shape
- Both Faces Are Reflective
- Plated Metal or Glass Construction
They have a circular shape and are made of plated metal or glass without overpainting, so both faces can be used as a mirror.
The focal length has the same value but opposite sign for both sides.
Spécifications
- Désignation:Convex/concave mirrors, glass
- Focal Length:200 mm
- Diameter:75 mm