Convex Flask Brushes | Avantor
Convex Flask Brushes
Supplier:   Mill-Rose
Convex Flask Brushes
Supplier:  Mill-Rose
Certificates
Convex Flask Brushes
Convex Flask Brushes
2 Options Available Below
Product Details & Documents
White hog bristles mounted on a durable, plastic handle. Flexible brush section adapts to curvature of flasks.
Product Family Options
Informations sur le produit
  • Désignation
    Bristle Material
    Bristle Color
    Handle Material
    Longueur totale
    Availability
    Price
  • Convex Flask Brushes
    Hog hair
    White
    Plastic
    305 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13.65
    $13.65
    Each
       $13.65
    Supplier Number: 2130
  • Convex Flask Brushes
    Hog hair
    White
    Plastic
    356 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10.75
    $10.75
    Each
       $10.75
    Supplier Number: 2131