Contour Model | Avantor
Contour Model
Supplier:   Science First
Contour Model
Supplier:  Science First
Certificates
Contour Model
3 Options Available Below
Product Details & Documents
Learn to draw contour lines and how to indicate elevations on a chart or map.
  • Visually connect 2D topo lines to 3D formations
    Hands-on interactive learning activity
    Activity can be repeated from class to class
    Single or class set available
The Contour Model Kit offers a hands-on approach to studying topography, terrain concepts, and landforms. By immersion in water, the model of Mount Capulin allows the shape of earth surface features to be displayed in two dimensions using hand drawn contour lines. Ideal for visual learning, this kit is great for demonstrating how topographic maps represent the natural features of the earth. Perfect for earth science and geography classrooms, this kit includes everything needed to build and study contour models, providing students with a practical understanding of landscapes and elevation changes.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • Contour model
    1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $69.35
    $69.35
    Each
       $69.35
    Supplier Number: 32200
  • Contour model (pack of 15)
    15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 15
    Pack of 15 - $507.20
    $507.20
    Pack of 15
       $507.20
    Supplier Number: 32210
  • Replacement case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.85
    $34.85
    Each
       $34.85
    Supplier Number: 32200-01