Conductivity/TDS Calibration Solution
Supplier: LaMotte
Jump to:
À propos de cet article
Always keep your meter in range.
- Already prepared solutions
- Various ranges
- Aligned with GLOBE protocols
Prepared solution to keep your conductivity calibrated. 500ml bottles.
3 Options Available Below
Product Details & Documents
Always keep your meter in range.
- Already prepared solutions
- Various ranges
- Aligned with GLOBE protocols
Prepared solution to keep your conductivity calibrated. 500ml bottles.
Product Family Options
Informations sur le produit
- DésignationConductivityDissolved SolidsAvailabilityPrice
- Supplier Number: 6416-L
- Supplier Number: 6417-L
- Supplier Number: 6354-L