Comprehensive Histology Slide Set
Comprehensive histology slide set, 205
Catalog # 470336-068
Supplier: Avantor
Jump to:
Product Details & Documents
The comprehensive histology slide set includes twelve distinct system sets.
- All identifying features are distinguishable
- Rigorous quality control standards
- In-House experts available to answer questions
Researchers investigate a variety of tissue and organ systems with this extensive histological slide set. All identifying features are clearly distinguishable, and the set meets rigorous quality control standards.
The slide set contents include general cytology (25 slides), epithelial tissues (five slides), general connective tissues (25 slides), general muscle tissues (10 slides), general nerve tissues (35 slides), vascular system (23 slides), endocrine system (four slides), digestive system (35 slides), respiratory system (seven slides), skin and appendages (nine slides), excretory system (six slides), and reproductive system (21 slides).
Spécifications
- Désignation:Comprehensive histology slide set
- No. of Slides:205