Compound Hand Magnifier
Compound Hand Magnifier, 2×, 6×, 8×
Catalog # 470105-656
Supplier: United Scientific Supplies
Fournisseur #: PMT003
Jump to:
Product Details & Documents
Two or more magnifiers may be snapped together to produce increased magnification and to illustrate the principles of compound lenses.
- Three magnifiers in one
- All plastic
Size: 100 mm.
Delivery information: Each magnifier includes 2x, 6x, and 8x lenses constructed of optical grade plastic.
Spécifications
- Grossissement:2×, 6×, 8×
- Désignation:Compound Hand Magnifier