Complete Precision Air Table | Avantor
Complete Precision Air Table
Supplier:   S R P INC CP
Complete Precision Air Table
Supplier:  S R P INC CP
Certificates
Complete Precision Air Table
Complete Precision Air Table
3 Options Available Below
Product Details & Documents
The Complete Precision Air Table helps investigate low-friction collisions.
  • Instructions with Suggested Experiments Included
  • Air Table Made of Heavy Glass
  • Air Compressor Included
  • Spark Sources Works with Six Frequencies
The recording system on this air table makes it easy to take good quantitative measurements from an air-table experiment.
A spark from the spark generator leaves a mark on the recording paper placed in the table. Students then use a protractor and centimeter scale to take measurements. The spark source can be operated at six different frequencies.
Storage recommendation for Carbon paper: Carbon papers should be stored in a dry place as the backing paper is sensitive to dampness. If not stored properly, the backing sheet will be difficult to remove from the carbon sheet.
Certifications: CSA approved.
Delivery information: This kit includes an air table made of heavy glass with a metal frame, air compressor, aluminum support for air hoses, air-supply hose, spark generator, two 550 g steel pucks, one 160 g auxiliary mass for unequal mass collisions, Velcro adhesive puck collars, two springs, four table pins, one orbital post with suction cup, two wire collars with hooks for pucks, a pulley, 100 sheets of recording paper, one sheet of carbon paper, and an instruction manual with suggested experiments.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Complete Precision Air Table
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,248.15
    $6,248.15
    Each
       $6,248.15
    Supplier Number: T01-001F
  • Carbon Paper, Single Sheet
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.20
    $31.20
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $1,484.20
    $1,484.20
    Each
       $31.20
    Pack of 50
       $1,484.20
    Supplier Number: F15-012
  • Recording Paper Sheets
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $142.75
    $142.75
    Pack of 100
       $142.75
    Supplier Number: 0011068
  • Recording Paper Sheets, Package of 100
    Discontinued
    Supplier Number: -
  • Complete Precision Air Table
    Discontinued
    Supplier Number: -
  • Carbon Paper, Single Sheet
    Discontinued
    Supplier Number: -