Columnar Epithelium, Isolated Cells Slide
Catalog # 470182-602
Supplier: Avantor
Specifications
- Description:Columnar Epithelium, Isolated Cells
- Cat. no.:470182-602
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-602
- Orientation:Smear
Specifications
About this item
Aids in Absorption and Mucus Movement
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Mucosa of small intestine. Maceration preparation.