Colored Pencils
Assorted, 3.3 mm, Colored pencils, set of 12
Catalog # 470351-422
Supplier: DIXON TICONDEROGA COMPANY
Fournisseur #: X22120X
Jump to:
Product Details & Documents
Pencil set with 12 assorted colors.
- Excellent color vibrancy
- Pre-sharpened
- Non-toxic
- 12 colors: White, orange, rose, brown, light blue, blue, violet, black, yellow, light green, dark green, and red
Length of pencil: 7"
Spécifications
- Lead Diameter:3.3 mm
- Color:Assorted
- Désignation:Colored pencils, set of 12