Coleus, Stem Tip Slide
Coleus, Stem Tip Slide, Longitudinal section, Quadruple
470181-316
: Avantor
The Coleus Stem Tip Slide demonstrates leaf development.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows apical meristem and developing leaves.
- Désignation:Coleus, Stem Tip Slide
- Stain:Quadruple
- Orientation:Longitudinal section