Clonorchis sinensis Slide
Clonorchis sinensis, Section, Hematoxylin and triosin
Catalog # 470181-982
Supplier: Avantor
Jump to:
Product Details & Documents
Human Parasite
- Identifying features clearly distinguishable
- Rigorous quality control standards
In situ from the liver. Feeds on bile.
Spécifications
- Désignation:Clonorchis sinensis
- Stain:Hematoxylin and triosin
- Orientation:Section