Cladophora
Cladophora (Small), Erdschreiber's medium, 30, Tube
Catalog # 470176-496
Supplier: Ward's Science
Supplier #: 152105
Jump to:
Product Details & Documents
Marine. Filamentous; freely branched. Erdschreiber’s medium.
Characterized by green plastids and cell walls composed of cellulose.
Spécifications
- Désignation:Cladophora (Small)
- No. of Students:30
- Packaging:Tube
- Media:Erdschreiber's medium