Citrus Cleaner
Catalogue # 470006-770
Fournisseur: Ward's Science
Spécifications
- Description:Citrus Cleaner
- Cat. no.:470006-770
- Supplier No.:CC0343-250
Spécifications
À propos de cet article
Squeeze bottle cleans grease, marker, adhesive, and wax from any surface.
Oil based cleaner not intended for use with lenses. Recommended by microscope repair specialist Joyce Dorsey for cleaning microscope bodies. 250 mL bottle.