The Circulatory System Microslide
The Circulatory System Microslide
Certificates
The Circulatory System Microslide
The Circulatory System Microslide
2 Options Available Below
Product Details & Documents
Explore the pathways and tissues of the circulatory system.
  • Arrows Help Students Locate Important Features
  • Mounted in a Clear Plastic Holder That Protects It on Both Sides
  • Set of 15 Includes Detailed Lesson Plan on the Topic Under Study
  • Durable, Inexpensive Teaching Resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions.
You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
 : This set includes human blood (sm, st, 900X), phagocyte (em, 25,000X), artery and vein (cs, st, 35X), capillaries in the lung (cs, i, 360X), lymph vessel and valves (wm, st, 60X), rat heart (ls, st, 4X), rat heart (cs, st, 6X), and valve at aorta (ls, st, 7 1/2X).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • The circulatory system microslide
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $19.60
    $19.60
    Pack of 1
       $19.60
    Supplier Number: T-68
  • The circulatory system microslides, set of 15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 15
    Box of 15 - $291.85
    $291.85
    Box of 15
       $291.85
    Supplier Number: T-68-15B