Chromatography Paper
46 x 57 cm (18 x 22"), 130 mm/30 min
Catalog # 470003-724
Supplier: AHLSTROM MUNKSJO FILTRATION LLC.
Fournisseur #: 6130-4657
Jump to:
Product Details & Documents
Quality paper for general chromatography and electrophoresis.
- White, smooth sheets with a hard surface
Spécifications
- Débit:130 mm/30 min
- Taille:46 x 57 cm (18 x 22")